Skip to Content
Merck
All Photos(1)

Key Documents

579211

Sigma-Aldrich

Tetrakis(dimethylamido)zirconium(IV)

electronic grade, ≥99.99% trace metals basis

Synonym(s):

TDMAZ, Tetrakis(dimethylamino)zirconium(IV)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2N]4Zr
CAS Number:
Molecular Weight:
267.53
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

grade

electronic grade

Assay

≥99.99% trace metals basis

form

solid

reaction suitability

core: zirconium

mp

57-60 °C (lit.)

storage temp.

2-8°C

SMILES string

CN(C)[Zr](N(C)C)(N(C)C)N(C)C

InChI

1S/4C2H6N.Zr/c4*1-3-2;/h4*1-2H3;/q4*-1;+4

InChI key

DWCMDRNGBIZOQL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Tetrakis(dimethylamido) zirconium (IV) may be used as a precursor for atomic layer deposition of zirconium which find applications ranging from gas sensors to high-k dielectrics in microelectronics.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3 - Water-react 2

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Atomic layer deposition of ZrO2 and HfO2 nanotubes by template replication
Gu D, et al.
Electrochemical and Solid-State Letters, 12.4, K25-K28 (2009)
Surface morphology and crystallinity control in the atomic layer deposition (ALD) of hafnium and zirconium oxide thin films.
Hausmann DM & Gordon RG
Journal of Crystal Growth, 249.1, 251-261 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service