Skip to Content
Merck
All Photos(1)

Key Documents

579033

Sigma-Aldrich

Disperse Yellow 7 methacrylate

95%

Synonym(s):

[2-methyl-4-[(4-phenyldiazenylphenyl)diazenyl]phenyl] 2-methylprop-2-enoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5N=NC6H4N=NC6H3(CH3)OCOC(CH3)=CH2
CAS Number:
Molecular Weight:
384.43
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

95%

form

solid

mp

132.7-135.8 °C (lit.)

λmax

364 nm

storage temp.

2-8°C

SMILES string

CC(=C)C(=O)Oc1ccc(cc1C)\N=N\c2ccc(cc2)\N=N\c3ccccc3

InChI

1S/C23H20N4O2/c1-16(2)23(28)29-22-14-13-21(15-17(22)3)27-26-20-11-9-19(10-12-20)25-24-18-7-5-4-6-8-18/h4-15H,1H2,2-3H3/b25-24+,27-26+

InChI key

NLSOMFRHHRRNDW-JQRDNDPDSA-N

General description

NLO material

Application

  • Synthesis and Characterization of Functional Thermoresponsive Photoactive Copolymers: This study explores the synthesis of a photoactive copolymer using Disperse Yellow 7 methacrylate, focusing on its thermal and photoluminescence properties. The research aims to enhance the applications of these materials in smart and responsive textiles (K Meenu, DS Bag, 2023).

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service