576697
Triethyl 1,3,5-triazine-2,4,6-tricarboxylate
97%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
Assay
97%
mp
170-174 °C (lit.)
functional group
ester
SMILES string
CCOC(=O)c1nc(nc(n1)C(=O)OCC)C(=O)OCC
InChI
1S/C12H15N3O6/c1-4-19-10(16)7-13-8(11(17)20-5-2)15-9(14-7)12(18)21-6-3/h4-6H2,1-3H3
InChI key
LKNPNZKIOSOWES-UHFFFAOYSA-N
Related Categories
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service