Skip to Content
Merck
All Photos(2)

Key Documents

559792

Sigma-Aldrich

[3-(3,5-Dichlorophenyl)-2,4-dioxoimidazolidinyl]-N-(methylethyl)carboxamide

97%

Synonym(s):

Iprodione

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3N2C3H2O2CONHCH(CH3)2
CAS Number:
Molecular Weight:
330.17
Beilstein:
895003
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

130-134 °C (lit.)

SMILES string

CC(C)NC(=O)N1CC(=O)N(C1=O)c2cc(Cl)cc(Cl)c2

InChI

1S/C13H13Cl2N3O3/c1-7(2)16-12(20)17-6-11(19)18(13(17)21)10-4-8(14)3-9(15)5-10/h3-5,7H,6H2,1-2H3,(H,16,20)

InChI key

ONUFESLQCSAYKA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jorge Morales et al.
Chemosphere, 92(11), 1536-1541 (2013-05-21)
The influence of humic aggregates in water solution upon the chemical stability of Iprodione has been investigated under basic conditions. Taking into account that an important part of soils are colloids, the possibility of its presence implies that soil composition
Osamu Maeda et al.
Journal of agricultural and food chemistry, 58(3), 1416-1419 (2009-12-17)
Iprodione is an agricultural fungicide that is difficult to detect in foods by HPLC because it coelutes with natural compounds in the food. We previously showed that food matrix could be degraded with titanium dioxide powder (TP). Here we describe
Leiyan Yan et al.
Fungal genetics and biology : FG & B, 47(9), 753-760 (2010-07-03)
We present a characterization of bos5 from Botrytis cinerea, a gene that encodes a mitogen-activated protein kinase kinase (MAPKK), which is homologous to OS-5 of Neurospora crassa. The bos5 gene deletion mutant exhibited reduced vegetative growth and strongly impaired conidiation.
Alexander B Fialkov et al.
Journal of chromatography. A, 1218(52), 9375-9383 (2011-11-29)
A new type of low thermal mass (LTM) fast gas chromatograph (GC) was designed and operated in combination with gas chromatography mass spectrometry (GC-MS) with supersonic molecular beams (SMB), including GC-MS-MS with SMB, thereby providing a novel combination with unique
Chad R Blystone et al.
Toxicology letters, 174(1-3), 74-81 (2007-10-13)
Iprodione (IPRO) is a dichlorophenyl dicarboximide fungicide similar to procymidone and vinclozolin. All three of these fungicides induce Leydig cell tumors in the rat testis in long-term studies and an endocrine mode of action has been hypothesized to mediate this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service