556459
Polyaniline (emeraldine base)
average Mw ~5,000
Synonym(s):
Emeraldine base polyaniline
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
CAS Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
mol wt
average Mw ~5,000
Quality Level
solubility
m-cresol: soluble
DMAC: soluble
DMF: soluble
DMSO: soluble
NMP: soluble
λmax
~325 nm
SMILES string
Brc1nn[nH]c1C.Cl
InChI
1S/C3H4BrN3.ClH/c1-2-3(4)6-7-5-2;/h1H3,(H,5,6,7);1H
InChI key
LYCWKZFKTMHXQA-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Magnetic susceptibility of emeraldine base polyaniline was reported.
Polyaniline (emeraldine base) (PANI) is anintrinsic conductive polymer which consists of a ring structure attached with nitrogen substituted derivatives. It has an emeraldine type oxidation state that exists in the form of its base. It can be prepared by deprotonation of emeraldine hydrochloric acid and ammonium hydroxide. It is mainly used in the development of electrochemical devices.
Application
Emeraldine base polyaniline may be used as a corrosion protection undercoat on steel and iron. It may be used as a hole transporting layer in polymer light emitting diodes.
PANI forms a conjugating system that can be used in semiconducting applications. It can be used in a variety of other applications which include protective coating, hole transporting layer for organic electronics and chemical sensing.
Features and Benefits
Undoped form of conducting polymer (non-conductive).
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Corrosion protection of iron/steel by emeraldine base polyaniline: an X-ray photoelectron spectroscopy study
Fahlman M, et al.
Synt. Metals, 85(1-3), 1323-1326 (1997)
A magnetic susceptibility study of emeraldine base polyaniline`
Kahol PK, et al.
Synt. Metals, 140(2-3), 261-267 (2004)
Suitability of emeraldine base polyaniline-PVA composite film for carbon dioxide sensing.
Irimia-Vladu M and Fergus JW
Synthetic Metals, 156(21-24), 1401-1407 (2006)
Corrosion protection of iron/steel by emeraldine base polyaniline: an X-ray photoelectron spectroscopy study.
Fahlman M, et al.
Synthetic Metals, 85(1-3), 1323-1326 (1997)
Application of polyaniline (emeraldine base, EB) in polymer light-emitting devices.
Wang HL, et al.
Synthetic Metals, 78(1), 33-37 (1996)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service