Skip to Content
Merck
All Photos(1)

Key Documents

554324

Sigma-Aldrich

Phenylacetyl disulfide

96%

Synonym(s):

Bis(phenylacetyl) disulfide, Di(phenylacetyl) disulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5CH2COS)2
CAS Number:
Molecular Weight:
302.41
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

solid

mp

59-63 °C (lit.)

functional group

disulfide
phenyl

SMILES string

O=C(Cc1ccccc1)SSC(=O)Cc2ccccc2

InChI

1S/C16H14O2S2/c17-15(11-13-7-3-1-4-8-13)19-20-16(18)12-14-9-5-2-6-10-14/h1-10H,11-12H2

InChI key

IXGZXXBJSZISOO-UHFFFAOYSA-N

General description

Phenylacetyl disulphide serves as a sulfurization reagent during the preparation of phosphates.Phenyl acetyl disulfide can be synthesized by the reaction of phenyl benzenethiolsulfonate with thioacetic acid in the presence of triethylamine.

Application

Phenylacetyl disulfide (PADS) may be used as a sulphur transfer agent during the synthesis of phosphorothioate oligodeoxyribonucleotides.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andreas Aemissegger et al.
Tetrahedron, 63(27), 6185-6190 (2007-07-02)
The synthesis of 1-(2-nitrophenylethyl) caged O-phosphorothioylserine, -threonine and -tyrosine derivatives is reported. These amino acid building blocks can be directly incorporated into peptides by Fmoc-based solid phase synthesis as their pentafluorophenyl esters or as symmetric anhydrides. Upon irradiation with UV
Use of phenylacetyl disulfide (PADS) in the synthesis of oligodeoxyribonucleotide phosphorothioates.
Cheruvallath ZS, et al.
Nucleosides, nucleotides & nucleic acids, 18(3), 485-492 (1999)
A study on the use of phenylacetyl disulfide in the solid-phase synthesis of oligodeoxynucleoside phosphorothioates.
Roelen HCPF, et al.
J. R. Neth. Chem. Soc., 110(7-8), 325-331 (1991)
"Organic disulfides and related substances. XXIII. Unsymmetrical carbonyl disulfides and cognate compounds"
Fiel L and Buckman DJ
The Journal of Organic Chemistry, 32(11), 3467-3470 (1967)
Synthesis of antisense oligonucleotides: Replacement of 3H-1, 2-benzodithiol-3-one 1, 1-dioxide (Beaucage reagent) with phenylacetyl disulfide (PADS) as efficient sulfurization reagent: From bench to bulk manufacture of active pharmaceutical ingredient.
Cheruvallath ZS, et al.
Organic Process Research & Development, 4(3), 199-204 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service