Skip to Content
Merck
All Photos(3)

Key Documents

553719

Sigma-Aldrich

2-Amino-3-bromopyridine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5BrN2
CAS Number:
Molecular Weight:
173.01
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

63-67 °C

functional group

bromo

SMILES string

Nc1ncccc1Br

InChI

1S/C5H5BrN2/c6-4-2-1-3-8-5(4)7/h1-3H,(H2,7,8)

InChI key

RBCARPJOEUEZLS-UHFFFAOYSA-N

Application

2-Amino-3-bromopyridine may be used to synthesize:
  • 2-acylamido-3-bromopyridines
  • 2-anilino-3-bromopyridine
  • 3-[(2-methoxyphenyl)ethynyl]pyridin-2-amine
  • 3-[(4-methoxyphenyl)ethynyl]pyridin-2-amine
  • N-(bromopyridyl)amidines
  • carbolines via palladium catalyzed arylation followed by palladium catalyzed amination reaction
  • 2-amino-3-cyanopyridine via palladium catalyzed cyanation reaction with potassium ferro-cyanide in the presence of 1,8-diazabicyclo[5.4.0]undec-7-ene [DBU]
  • nitro-substituted N,N′-dipyridinylamines via palladium catalyzed coupling reaction with 2-chloro-3-nitropyridine in the presence of Xantphos ligand
  • 2-amino-3-iodopyridine via reaction with sodium iodide in the presence of copper(I)iodide and trans-N,N′-dimethylcyclohexane-1,2-diamine

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Syntheses of pyrido [1, 2-a][1, 3, 5] triazin-4-one C-deoxyribonucleosides"
Oda H, et al.
Tetrahedron, 64(45), 11021-11029 (2007)
Novel synthetic strategy of carbolines via palladium-catalyzed amination and arylation reaction.
Iwaki T, et al.
Journal of the Chemical Society. Perkin Transactions 1, 11, 1505-1510 (1999)
Synthesis of pyridopyrimidines by palladium-catalyzed isocyanide insertion.
Este'vez V, et al.
ACS Catalysis, 4(1), 40-43 (2013)
"Smiles Rearrangement as a Tool for the Preparation of Dihydrodipyridopyrazines"
Patriciu I-O, et al.
Organic Letters, 11(23), 5502-5505 (2009)
Phosphonylation of 2-Amino-and 2-Amido-3-bromopyridines and 2-Amino-3-chloroquinoxalines with Triethyl Phosphite.
Adam M, et al.
European Journal of Organic Chemistry, 27, 4655-4665 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service