Skip to Content
Merck
All Photos(1)

Documents

529176

Sigma-Aldrich

3,4,5-Trifluorobenzaldehyde

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F3C6H2CHO
CAS Number:
Molecular Weight:
160.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

impurities

≤3%

refractive index

n20/D 1.482 (lit.)

bp

174 °C (lit.)

density

1.42 g/mL at 25 °C (lit.)

SMILES string

Fc1cc(C=O)cc(F)c1F

InChI

1S/C7H3F3O/c8-5-1-4(3-11)2-6(9)7(5)10/h1-3H

InChI key

NLAVHUUABUFSIG-UHFFFAOYSA-N

Application

3,4,5-Trifluorobenzaldehyde may be used in the synthesis of 3,4,5-tris(methylthio)benzaldehyde.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Repr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kunhao Li et al.
Inorganic chemistry, 46(12), 4844-4849 (2007-05-10)
We introduce the 1,2,3-tris(organylthiophenyl) group as a symmetrical, multidentate chelation link for building coordination networks. For this, zinc(II) 5,10,15,20-tetrakis[3',4',5'-tris(methylthio)phenyl]porphyrin was synthesized and integrated into a two-dimensional network via coordination with BiBr3. The coordination link exhibits an unusually complex bonding pattern
James P Hagen et al.
Bioorganic chemistry, 97, 103590-103590 (2020-03-18)
Insect-borne parasite Trypanosoma brucei plagues humans and other animals, eliciting the disease Human African trypanosomiasis, also known as African sleeping sickness. This disease poses the biggest threat to the people in Sub-Saharan Africa. Given the high toxicity and difficulties with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service