Skip to Content
Merck
All Photos(2)

Key Documents

52865

Sigma-Aldrich

Sodium 1-hexanesulfonate monohydrate

≥98.0% (T)

Synonym(s):

1-Hexanesulfonic acid sodium salt monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH2SO3Na · H2O
CAS Number:
Molecular Weight:
206.24
Beilstein:
3727014
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (T)

form

solid

functional group

sulfonic acid

SMILES string

O.[Na+].CCCCCCS([O-])(=O)=O

InChI

1S/C6H14O3S.Na.H2O/c1-2-3-4-5-6-10(7,8)9;;/h2-6H2,1H3,(H,7,8,9);;1H2/q;+1;/p-1

InChI key

CLCGFJYKZGFGSQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium 1-pentanesulfonate monohydrate, also known as 1-Hexanesulfonic acid sodium salt monohydrate, is commonly used as a surfactant and a reversed-phase ion-pairing agent.

Application

Sodium 1-hexanesulfonate monohydrate can be used as a catalyst to synthesize:     
  • Amidoalkyl naphthols by three-component reaction of β-naphthol, aldehydes, and urea.      
  • Aminophosphonates by the reaction of aldehydes/ketones with amines.

It can also be used as a reactant to prepare polynuclear oxidobismuth sulfonates by reacting with sodium 2-methyl-2-propensulfonate and tetra-μ3-hydroxyhexakis(nitrato-κO)tetra-μ3-oxohexabismuth in DMSO.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid arsenic speciation using ion pair LC-ICPMS with a monolithic silica column reveals increased urinary DMA excretion after ingestion of rice
Pearson G, et al.
Journal of Analytical Atomic Spectrometry, 22(4), 361-369 (2007)
Kirti S Niralwad et al.
Ultrasonics sonochemistry, 17(5), 760-763 (2010-03-17)
1-Hexanesulphonic acid sodium salt was found to be an efficient catalyst for the green synthesis of alpha-aminophosphonates by the coupling of aldehydes/ketone, an amine and triethyl phosphite under ultrasound irradiation at ambient temperature for appropriate time to furnish the desired
Philip Zakaria et al.
Electrophoresis, 23(17), 2821-2832 (2002-09-11)
The separation of a series of aromatic carboxylic acids, sulfonates and opiates using electrokinetic chromatography employing a mixture of the soluble cationic polymer poly(diallydimethylammonium chloride) (PDDAC) and the amphiphilic anion hexanesulfonate as pseudostationary phases is described. In this system, the
A R Calhoun et al.
Journal of colloid and interface science, 309(2), 505-510 (2007-03-03)
Measurements have been made to determine the solubility of ethane, C2H6, in aqueous solutions of four different surfactants of the linear alkanesulfonate class at 25 degrees C. The surfactants, sodium 1-pentanesulfonate, sodium 1-hexanesulfonate, sodium 1-heptanesulfonate, and sodium 1-octanesulfonate, all share
Ana L Costa et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(34), 12069-12078 (2015-07-29)
Zn-Al layered double hydroxides (LDHs) containing solely indigo carmine (IC) or 1-hexanesulfonate (HS) anions, or a mixture of the two with different HS/IC molar ratios, were prepared by the direct synthesis method and characterized by various techniques. Hydrotalcite-type phases were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service