Skip to Content
Merck
All Photos(2)

Documents

519596

Sigma-Aldrich

4-Amino-3-chlorobenzonitrile

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H3(Cl)CN
CAS Number:
Molecular Weight:
152.58
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

102-105 °C (lit.)

SMILES string

Nc1ccc(cc1Cl)C#N

InChI

1S/C7H5ClN2/c8-6-3-5(4-9)1-2-7(6)10/h1-3H,10H2

InChI key

OREVCMGFYSUYPX-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Tsuji et al.
Chemical & pharmaceutical bulletin, 40(9), 2399-2409 (1992-09-01)
Various 2'-phenoxymethanesulfonanilide derivatives were synthesized and evaluated for antiinflammatory and analgesic activities. Some compounds bearing an electron-attracting substituent at the 4'-position strongly inhibited adjuvant-induced arthritis in rats and acetic acid-induced writhing syndrome in mice without causing gastro-intestinal irritation. Among them
Australian Journal of Chemistry, 35, 328-328 (1982)
Crystal structures of Pigment Red 170 and derivatives, as determined by X-ray powder diffraction.
Martin U Schmidt et al.
Angewandte Chemie (International ed. in English), 45(8), 1313-1317 (2006-01-21)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service