Skip to Content
Merck
All Photos(1)

Key Documents

51682

Sigma-Aldrich

1-Ethyl-3-methylimidazolium ethyl sulfate

≥95%

Synonym(s):

[EMIM][ESO4]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H16N2O4S
CAS Number:
Molecular Weight:
236.29
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95%

SMILES string

CCOS([O-])(=O)=O.CCn1cc[n+](C)c1

InChI

1S/C6H11N2.C2H6O4S/c1-3-8-5-4-7(2)6-8;1-2-6-7(3,4)5/h4-6H,3H2,1-2H3;2H2,1H3,(H,3,4,5)/q+1;/p-1

InChI key

VRFOKYHDLYBVAL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

1-Ethyl-3-methylimidazolium ethyl sulfate is a water-miscible, aprotic room temperature ionic liquid (RTIL). It can be prepared by reacting 1-methylimidazole with diethyl sulfate.

Application

[EMIM][ESO4] has been used to study its effect:
  • As a solvent on the thermal stability of lysozyme.
  • As an additive on potassium hydroxide (KOH) electrolyte properties and hydrogen evolution reaction (HER) reaction kinetics.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Muta. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

310.1 °F

Flash Point(C)

154.5 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solubility and diffusion of CO 2 and H 2 S in the ionic liquid 1-ethyl-3-methylimidazolium ethylsulfate
Jalili, Amir Hossein, et al.
The Journal of Chemical Thermodynamics, 42.10, 1298-1303 (2010)
Experimental study of the density and viscosity of 1-ethyl-3-methylimidazolium ethyl sulfate
Schmidt, H., et al.
The Journal of Chemical Thermodynamics, 47, 68-75 (2012)
Electronic structure and normal vibrations of the 1-ethyl-3-methylimidazolium ethyl sulfate ion pair
Dhumal, Nilesh R., Hyung J. Kim, and Johannes Kiefer
The Journal of Physical Chemistry A, 115.15, 3551-3558 (2011)
Impact of imidazolium-based ionic liquids on the structure and stability of lysozyme.
Satish L, et al.
Spectroscopy Letters, 49(6), 383-390 (2016)
Surface studies on the ionic liquid 1-ethyl-3-methylimidazolium ethylsulfate using X-ray photoelectron spectroscopy (XPS)
Gottfried, J. M., et al.
Zeitschrift fur Physikalische Chemie (Frankfurt am Main, Germany), 220.10, 1439-1453 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service