493325
Sodium propionate-1,2-13C2
99 atom % 13C
Synonym(s):
Propionic acid-1,2-13C2 sodium salt
About This Item
Recommended Products
isotopic purity
99 atom % 13C
form
solid
technique(s)
mass spectrometry (MS): suitable
mp
285-286 °C (lit.)
mass shift
M+2
SMILES string
[Na+].C[13CH2][13C]([O-])=O
InChI
1S/C3H6O2.Na/c1-2-3(4)5;/h2H2,1H3,(H,4,5);/q;+1/p-1/i2+1,3+1;
InChI key
JXKPEJDQGNYQSM-MEFQWSPQSA-M
Packaging
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service