481696
Poly(dimethylsiloxane), bis(3-aminopropyl) terminated
average Mn ~27,000
Synonym(s):
PDMS
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
specific activity
0.07-0.09 meq/g (Amine number)
Quality Level
mol wt
average Mn ~27,000
refractive index
n20/D 1.406
viscosity
2,000 cSt
bp
>200 °C (lit.)
density
0.98 g/mL at 25 °C (lit.)
SMILES string
C[Si](C)=O.C[Si](C)(O)CCCN
InChI
1S/C5H15NOSi.C2H6OSi/c1-8(2,7)5-3-4-6;1-4(2)3/h7H,3-6H2,1-2H3;1-2H3
InChI key
LYVFZZVBTQSOBI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
- Synthesis and Characterization of Self-Healing Elastomers Based on Coordination Bonds: This study explores the use of Aminopropyl terminated polydimethylsiloxane (APT-PDMS) in creating self-healing elastomers, highlighting its potential in materials engineering and design (BAI Zhiwei et al. 2020).
- Poly (imide-siloxane)s based on hyperbranched polyimides: Discusses the synthesis of polyimide-siloxane copolymers using bis(3-aminopropyl)-terminated poly(dimethylsiloxane), illustrating the versatility of this material in creating advanced polymer structures (P Sysel et al., 2018).
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service