Skip to Content
Merck
All Photos(3)

Key Documents

480118

Sigma-Aldrich

3,4-Dimethoxyphenylboronic acid

≥95.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3B(OH)2
CAS Number:
Molecular Weight:
181.98
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95.0%

mp

245-250 °C (lit.)

SMILES string

COc1ccc(cc1OC)B(O)O

InChI

1S/C8H11BO4/c1-12-7-4-3-6(9(10)11)5-8(7)13-2/h3-5,10-11H,1-2H3

InChI key

RCVDPBFUMYUKPB-UHFFFAOYSA-N

Application

3,4-Dimethoxyphenylboronic acid can be used:
  • As a substrate in the cross-coupling reaction with 5,7-dichloropyrido[4,3-d]pyrimidine catalyzed by palladium.
  • As a starting material for the synthesis of buflavine 1, a natural alkaloid.
  • In one of the key synthetic steps for the preparation of lipidated malarial glycosylphosphatidylinositols (GPI) disaccharide.
  • To prepare 3,3″,4,4″-tetramethoxy-1,1′:4′,1″-terphenyl by reacting with 1,4-dibromobenzene using Pd catalyst.

Other Notes

Contains varying amounts of anhydride

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

3, 3″, 4, 4″-Tetramethoxy-1, 1′: 4′, 1″-terphenyl
Pui L, et al.
Acta Crystallographica Section E, Structure Reports Online, 67(Pt 8), o1892-o1892 (2011)
Highly efficient synthesis of buflavine: a unique Amaryllidaceae alkaloid
Sahakitpichan P and Ruchirawat S
Tetrahedron Letters, 44(28), 5239-5241 (2003)
Regioselective cross-coupling reactions and nucleophilic aromatic substitutions on a 5, 7-dichloropyrido [4, 3-d] pyrimidine scaffold
Jang M, et al.
Tetrahedron Letters, 47(50), 8917-8920 (2006)
A Suzuki-Miyaura coupling mediated deprotection as key to the synthesis of a fully lipidated malarial GPI disaccharide
Liu X and Seeberger PH
Chemical Communications (Cambridge, England), 15, 1708-1709 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service