Skip to Content
Merck
All Photos(1)

Documents

476234

Sigma-Aldrich

2-Allyloxyethanol

98%

Synonym(s):

Ethylene glycol allyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCH2OCH2CH2OH
CAS Number:
Molecular Weight:
102.13
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

refractive index

n20/D 1.436 (lit.)

bp

159 °C (lit.)

density

0.955 g/mL at 25 °C (lit.)

SMILES string

C=CCOCCO

InChI

1S/C5H10O2/c1-2-4-7-5-3-6/h2,6H,1,3-5H2

InChI key

GCYHRYNSUGLLMA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Technological Aspects of Synthesis of Poly (ethylene glycol) Mono-1-propenyl Ether Monomers: This study investigates the isomerization reactions of 2-allyloxyethanol among other compounds, aiming to optimize synthesis processes for industrial applications (M Urbala, Polish Journal of Chemical Technology, 2020).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

M A Pinigin et al.
Gigiena i sanitariia, (2)(2), 4-6 (1995-03-01)
The threshold irritable concentration of 2-allyl oxyethanol (2-AOE) in ambient air was determined at the level of 0.68 mg/m3. The threshold of chronic effect of 2-AOE is 0.075 mg/m3. The mean daily permissible concentration of 2-AOE is suggested at the
Zhongyu Cai et al.
ACS sensors, 2(10), 1474-1481 (2017-09-25)
Lectin proteins, such as the highly toxic lectin protein, ricin, and the immunochemically important lectin, jacalin, play significant roles in many biological functions. It is highly desirable to develop a simple but efficient method to selectively detect lectin proteins. Here
G M Morris et al.
Cell proliferation, 30(8-9), 311-323 (1997-08-01)
The daily topical application of two compounds, a cream containing 10% evening primrose oil (EPO) and Lioxasol (a compound used clinically to treat radiation burns), resulted in increased cell proliferative activity in the skin of female Large White pigs. The
Ji Hae Choi et al.
Contrast media & molecular imaging, 2018, 4617493-4617493 (2018-07-27)
Novel probe development for positron emission tomography (PET) is leading to expanding the scope of molecular imaging. To begin responding to challenges, several biomaterials such as natural products and small molecules, peptides, engineered proteins including affibodies, and antibodies have been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service