Skip to Content
Merck
All Photos(1)

Key Documents

476196

Sigma-Aldrich

Dibutyl itaconate

96%

Synonym(s):

Di(n -butyl) itaconate, Dibutyl itaconate, 2-Methylenebutanedioic acid1,4-dibutyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3O2CCH2C(=CH2)CO2(CH2)3CH3
CAS Number:
Molecular Weight:
242.31
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

96%

refractive index

n20/D 1.444 (lit.)

bp

284 °C (lit.)

density

0.985 g/mL at 25 °C (lit.)

SMILES string

CCCCOC(=O)CC(=C)C(=O)OCCCC

InChI

1S/C13H22O4/c1-4-6-8-16-12(14)10-11(3)13(15)17-9-7-5-2/h3-10H2,1-2H3

InChI key

OGVXYCDTRMDYOG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dibutyl itaconate (DBI) is an itaconate ester that can be prepared by an itaconic acid precursor.

Application

DBI is a monomer that can be copolymerized with pyridine groups to form nanocomposites for the development of bio-based elastomers. Copolymerization of DBI with acrylated epoxidized soya oil (AESO) can produce a thermoset material which finds potential applications in the modification of fatty acids.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

276.8 °F

Flash Point(C)

136 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Design and synthesis by redox polymerization of a bio-based carboxylic elastomer for green tire.
Zhou X, et al.
Science China: Chemistry, 58(10), 1561-1569 (2015)
Esterification of itaconic acid using Ln~ SO42-/TiO2-SiO2 (Ln= La3+, Ce4+, Sm3+) as catalysts.
Li Lu, et al.
J. Mol. Catal. A: Chem., 368, 24-30 (2013)
Design and preparation of natural layered silicate/bio-based elastomer nanocomposites with improved dispersion and interfacial interaction.
Qiao H, et al.
Polymer, 79, 1-11 (2015)
Properties of thermosets derived from chemically modified triglycerides and bio-based comonomers.
Beach ES, et al.
Applied Sciences, 3(4), 684-693 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service