Skip to Content
Merck
All Photos(3)

Key Documents

467871

Sigma-Aldrich

Sodium thiocyanate

≥99.99% trace metals basis

Synonym(s):

Sodium isothiocyanate, Sodium rhodanate, Sodium rhodanide, Sodium sulfocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaSCN
CAS Number:
Molecular Weight:
81.07
Beilstein:
3594965
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥99.99% trace metals basis

impurities

≤0.005% insolubles
≤0.2% Na2CO3
<100 ppm total metallic impurities

mp

287 °C (dec.) (lit.)

anion traces

S2-: ≤0.001%
chloride (Cl-): ≤0.01%
sulfate (SO42-): ≤0.01%

cation traces

Fe: ≤2 ppm
NH4+: ≤0.002%
heavy metals: ≤5 ppm

SMILES string

[Na]SC#N

InChI

1S/CHNS.Na/c2-1-3;/h3H;/q;+1/p-1

InChI key

VGTPCRGMBIAPIM-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Morphological and differential thermal analysis of complexes of sodium thiocyanate with poly(ethylene oxide) have been studied.[1] Ionic deep eutectic solvents composed of acetamide+lithium nitrate/sodium thiocyanate have been investigated by dielectric relaxation spectroscopy.[2]

Application

Sodium thiocyanate may be used as a reducing agent in the preparation of monodisperse gold oligoclusters.[3]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1

Supplementary Hazards

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Satya N Tripathy et al.
The Journal of chemical physics, 142(18), 184504-184504 (2015-05-17)
A detailed investigation on the molecular dynamics of ionic deep eutectic solvents (acetamide + lithium nitrate/sodium thiocyanate) is reported. The study was carried out employing dielectric relaxation spectroscopy covering seven decades in frequency (10(-1)-10(6) Hz) and in a wide temperature
Morphology and ionic conductivity of complexes of sodium iodide and sodium thiocyanate with poly (ethylene oxide).
Lee CC and Wright PV.
Polymer, 23(5), 681-689 (1982)
A Simple Method for the Size Controlled Synthesis of Stable Oligomeric Clusters of Gold Nanoparticles under Ambient Conditions.
Lawrence M, et al.
Journal of Visualized Experiments, 108, e53388-e53388 (2016)
Nancy E Breen et al.
PeerJ, 7, e6644-e6644 (2019-04-12)
The illegal practice of using cyanide (CN) as a stunning agent to collect fish for both the marine aquarium and live fish food trades has been used throughout the Indo-Pacific for over 50 years. CN fishing is destructive to all
S-F Chin et al.
Molecular pathology : MP, 56(5), 275-279 (2003-09-30)
To describe a robust pretreatment protocol for preparing paraffin wax embedded tissues on tissue microarrays for fluorescence in situ hybridisation (FISH). The newly developed pretreatment protocol described here was compared with the commonly used sodium thiocyanate based protocol and two

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service