Skip to Content
Merck
All Photos(1)

Key Documents

464384

Sigma-Aldrich

(S)-(−)-2-Hydroxy-4-(2-methoxyphenyl)-5,5-dimethyl-1,3,2-dioxaphosphorinane 2-oxide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17O5P
CAS Number:
Molecular Weight:
272.23
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

optical activity

[α]22/D −59°, c = 1 in methanol

mp

194-196 °C (lit.)

storage temp.

2-8°C

SMILES string

COc1ccccc1[C@H]2OP(O)(=O)OCC2(C)C

InChI

1S/C12H17O5P/c1-12(2)8-16-18(13,14)17-11(12)9-6-4-5-7-10(9)15-3/h4-7,11H,8H2,1-3H3,(H,13,14)/t11-/m1/s1

InChI key

HNFXKRNIAWHFJN-LLVKDONJSA-N

Application

Chiral phosphoric acid used for the resolution of amines, amino acids, and amino alcohols.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ten Hoeve, W. and Wynberg, H.
Synthetic Communications, 24, 899-899 (1994)
ten Hoeve, W., and Wynberg, H.
The Journal of Organic Chemistry, 50, 4508-4508 (1985)
A.D. van der Maest et al.
Rec. Trav. Chim., 109, 523-523 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service