463701
Manganese(III) oxide
99.9% trace metals basis
Synonym(s):
Manganese sesquioxide
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
Mn2O3
CAS Number:
Molecular Weight:
157.87
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Quality Level
Assay
99.9% trace metals basis
form
solid
density
4.5 g/mL at 25 °C (lit.)
application(s)
battery manufacturing
SMILES string
O=[Mn]O[Mn]=O
InChI
1S/2Mn.3O
InChI key
GEYXPJBPASPPLI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Manganese(III) oxide (Mn2O3) can be used for a variety of potential applications such as supercapacitors, fuel cells, zinc ion batteries, and lithium oxygen batteries.
Precursor to LiMn2O4. A useful battery cathode.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Manganese sesquioxide as cathode material for multivalent zinc ion battery with high capacity and long cycle life
Jiang B, et al.
Electrochimica Acta, 229(1), 422-428 (2017)
Alcohol oxidation on nanocrystalline oxide Pd/C promoted electrocatalysts
Shen PK and Xu C
Electrochemical Communications, 8(1), 184-188 (2006)
One-dimensional RuO2/Mn2O3 hollow architectures as efficient bifunctional catalysts for lithium-oxygen batteries
Yoon KR, et al.
Nano Letters, 16(3), 2076-2083 (2016)
Facile coating of manganese oxide on tin oxide nanowires with high-performance capacitive behavior
Yan J, et al.
ACS Nano, 4(7), 4247-4255 (2010)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service