Skip to Content
Merck
All Photos(1)

Key Documents

46210

Sigma-Aldrich

(+)-Fenchone

purum, ≥98.0% (sum of enantiomers, GC)

Synonym(s):

(+)-1,3,3-Trimethyl-2-norbornanone, (1S)-1,3,3-Trimethylbicyclo[2.2.1]heptan-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
Beilstein:
2206555
EC Number:
MDL number:
UNSPSC Code:
12352212
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

grade

purum

Quality Level

Assay

≥98.0% (sum of enantiomers, GC)

optical activity

[α]20/D +60±3°, neat

refractive index

n20/D 1.463

bp

63-65 °C/13 mmHg (lit.)

mp

5-7 °C (lit.)

density

0.945 g/mL at 20 °C (lit.)

functional group

ketone

SMILES string

CC1(C)[C@@H]2CC[C@@](C)(C2)C1=O

InChI

1S/C10H16O/c1-9(2)7-4-5-10(3,6-7)8(9)11/h7H,4-6H2,1-3H3/t7-,10+/m1/s1

InChI key

LHXDLQBQYFFVNW-XCBNKYQSSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giovanna Longhi et al.
The journal of physical chemistry. A, 110(15), 4958-4968 (2006-04-14)
We report and discuss the infrared (IR) vibrational circular dichroism (VCD) spectra of the enantiomeric pairs of the olefin derivatives of fenchone (1,3,3-trimethyl-2-methylenebicyclo[2.2.1]heptane) and camphor (1,7,7-trimethyl-2-methylenebicyclo[2.2.1]heptane), respectively, together with those of the parent molecules. The VCD spectra were taken in
B Simándi et al.
Journal of agricultural and food chemistry, 47(4), 1635-1640 (1999-11-24)
Ground fennel seeds were extracted with supercritical carbon dioxide. Small-scale subsequent extractions of the same sample showed that the composition of volatile compounds was changed with the extension of extraction time and only principal volatile components (limonene, fenchone, methylchavicol, and
I M Isa et al.
Talanta, 87, 230-234 (2011-11-22)
A new cobalt(II) ion selective electrode based on palladium(II) dichloro acetylthiophene fenchone azine(I) has been developed. The best membrane composition is found to be 10:60:10:21.1 (I)/PVC/NaTPB/DOP (w/w). The electrode exhibits a Nerstian response in the range of 1.0 × 10(-1)-1.0
Boris Steuer et al.
Phytochemical analysis : PCA, 14(5), 285-289 (2003-10-01)
The aim of this study was to investigate the accuracy and transferability of near-infrared (NIR) calibrations for estimating the content and composition of the volatile fraction in fennel fruits (Foeniculum vulgare Miller) as an example of medicinal and spice plants.
Dirk W Lachenmeier et al.
Journal of agricultural and food chemistry, 56(9), 3073-3081 (2008-04-19)
Thirteen samples of authentic absinthe dating from the preban era (i.e., prior to 1915) were analyzed for parameters that were hypothesized as contributing to the toxicity of the spirit, including naturally occurring herbal essences (thujone, pinocamphone, fenchone), methanol, higher alcohols

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service