Skip to Content
Merck
All Photos(1)

Key Documents

459798

Sigma-Aldrich

Allyl alcohol

≥98.5%

Synonym(s):

2-Propen-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH2OH
CAS Number:
Molecular Weight:
58.08
Beilstein:
605307
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.21

vapor density

2 (vs air)

vapor pressure

23.8 mmHg ( 25 °C)

Assay

≥98.5%

autoignition temp.

712 °F

expl. lim.

18 %

refractive index

n20/D 1.412 (lit.)

bp

96-98 °C (lit.)

mp

−129 °C (lit.)

density

0.854 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

OCC=C

InChI

1S/C3H6O/c1-2-3-4/h2,4H,1,3H2

InChI key

XXROGKLTLUQVRX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used to induce a mouse model of liver damage that has been used to study the mechanisms of hepatotoxicity and hepatic stem cell-mediated repair.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deoxydehydration of glycerol to allyl alcohol catalyzed by rhenium derivatives.
Canale V, et al.
Catalysis Science & Technology, 4(10), 3697-3704 (2014)
Hydrosilylation of allyl alcohol with [HSiMe2OSiO1.5]8: octa (3-hydroxypropyldimethylsiloxy) octasilsesquioxane and its octamethacrylate derivative as potential precursors to hybrid nanocomposites.
Zhang C and Laine RM.
Journal of the American Chemical Society, 122(29), 6979-6988 (2000)
Gregory M Mullen et al.
Journal of the American Chemical Society, 136(17), 6489-6498 (2014-04-08)
Partial oxidation of alcohols is a topic of great interest in the field of gold catalysis. In this work, we provide evidence that the partial oxidation of allyl alcohol to its corresponding aldehyde, acrolein, over oxygen-precovered gold surfaces occurs via
Madeleine C Warner et al.
Organic letters, 14(19), 5094-5097 (2012-09-26)
A general and efficient route for the synthesis of enantiomerically pure α-substituted ketones and the corresponding lactones has been developed. Ruthenium- and enzyme-catalyzed dynamic kinetic resolution (DKR) with a subsequent Cu-catalyzed α-allylic substitution are the key steps of the route.
Copper-catalyzed tandem trifluoromethylation/semipinacol rearrangement of allylic alcohols.
Zhi-Min Chen et al.
Angewandte Chemie (International ed. in English), 52(37), 9781-9785 (2013-07-23)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service