458937
Methyl (R)-(+)-3-Boc-2,2-dimethyl-4-oxazolidinecarboxylate
98%
Synonym(s):
Methyl (R)-(+)-3-(tert-butoxycarbonyl)-2,2-dimethyl-4-oxazolidinecarboxylate
About This Item
Recommended Products
Assay
98%
optical activity
[α]20/D +54°, c = 1.3 in chloroform
optical purity
ee: 99% (HPLC)
refractive index
n20/D 1.444 (lit.)
bp
89 °C/0.8 mmHg (lit.)
density
1.082 g/mL at 25 °C (lit.)
functional group
ester
ether
SMILES string
COC(=O)[C@H]1COC(C)(C)N1C(=O)OC(C)(C)C
InChI
1S/C12H21NO5/c1-11(2,3)18-10(15)13-8(9(14)16-6)7-17-12(13,4)5/h8H,7H2,1-6H3/t8-/m1/s1
InChI key
ZNBUXTFASGDVCL-MRVPVSSYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
199.4 °F - closed cup
Flash Point(C)
93 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service