Skip to Content
Merck
All Photos(2)

Key Documents

455830

Sigma-Aldrich

Ammonium hydrogen difluoride

99.999% trace metals basis

Synonym(s):

Ammonium bifluoride, Etching powder

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4HF2
CAS Number:
Molecular Weight:
57.04
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38050109
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.999% trace metals basis
form:
crystalline

Quality Level

Assay

99.999% trace metals basis

form

crystalline

impurities

≤15.0 ppm Trace Metal Analysis

mp

125 °C (lit.)

SMILES string

N.F.F

InChI

1S/2FH.H3N/h2*1H;1H3

InChI key

KVBCYCWRDBDGBG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Ammonium hydrogen difluoride is a white crystalline solid commonly used as an etchant, especially in the semiconductor industry. It has excellent selectivity in etching silicon oxides and metal silicides, making it useful in the manufacturing of semiconductors and microelectronic devices. It can also be used as a fluorinating agent.

Application

Used for the preparation of ammonium aminosilylfluorides and complex metal fluorides. Employed in a new hydrothermal synthesis of complex flourides NaHoF4 and NaEuF4; these interesting compounds have the fluorite structure and are important for solid-state lasers as well as for photoluminescence and magnetic characteristics

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zeitschrift fur Anorganische und Allgemeine Chemie, 622, 1646-1646 (1996)
Fluorination of sulfide minerals with ammonium hydrogen difluoride
N.M Laptash et al.
Journal of Fluorine Chemistry, 105, 53-58 (2000)
Chemical Communications (Cambridge, England), 1641-1641 (1996)
Acid etching as surface treatment method for luting of glass-ceramic restorations, part 1: acids, application protocol and etching effectiveness
Bajraktarova-Valjakova, et al.
Open access Macedonian journal of medical sciences, 6, 568-568 (2018)
Xun, X. et al.
Chemistry of Materials, 9, 2966-2966 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service