Skip to Content
Merck
All Photos(1)

Key Documents

452300

Sigma-Aldrich

Pyridinium trifluoromethanesulfonate

97%

Synonym(s):

Pyridinium triflate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6F3NO3S
CAS Number:
Molecular Weight:
229.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

221-223 °C (lit.)

SMILES string

c1ccncc1.OS(=O)(=O)C(F)(F)F

InChI

1S/C5H5N.CHF3O3S/c1-2-4-6-5-3-1;2-1(3,4)8(5,6)7/h1-5H;(H,5,6,7)

InChI key

YWVYZMVYXAVAKS-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of Lewis and Br?nsted acids on the homopolymerization of acrylates and their copolymerization with 1-alkenes.
Luo R, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 46(16), 5499-5505 (2008)
Caitlan E Ayala et al.
Angewandte Chemie (International ed. in English), 54(15), 4641-4645 (2015-02-20)
A new method which enables carbon-carbon bond formation at the α'-position of silylenol ethers by using catalytic amounts of pyridinium triflate is reported. This chemistry successfully produces, structurally challenging, highly substituted indole-containing silylenol ethers in excellent yields with complete regiocontrol
Dominik Jesariew et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 148, 203-214 (2015-04-19)
Infrared spectroscopy (4000-400 cm(-1)) in the wide temperature range, from 11 to 473 K, has been used to investigate the non-centrosymmetric pyridinium trifluoromethanesulfonate crystal, exhibiting several phase transitions. The assignments of the bands observed in the studied spectra have been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service