449776
Cobalt(II) chloride
AnhydroBeads™, −10 mesh, 99.9% trace metals basis
Synonym(s):
Cobalt dichloride, Cobalt muriate, Cobaltous chloride, Dichlorocobalt
About This Item
Recommended Products
grade
synthesis grade
Quality Level
vapor pressure
40 mmHg ( 0 °C)
product line
AnhydroBeads™
Assay
99.9% trace metals basis
impurities
≤1500.0 ppm Trace Metal Analysis
particle size
−10 mesh
mp
724 °C (lit.)
application(s)
battery manufacturing
SMILES string
Cl[Co]Cl
InChI
1S/2ClH.Co/h2*1H;/q;;+2/p-2
InChI key
GVPFVAHMJGGAJG-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Legal Information
accessory
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Eye Dam. 1 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service