Skip to Content
Merck
All Photos(2)

Key Documents

448958

Sigma-Aldrich

Thymine-1-acetic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8N2O4
CAS Number:
Molecular Weight:
184.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

272-278 °C (dec.) (lit.)

functional group

carboxylic acid

SMILES string

CC1=CN(CC(O)=O)C(=O)NC1=O

InChI

1S/C7H8N2O4/c1-4-2-9(3-5(10)11)7(13)8-6(4)12/h2H,3H2,1H3,(H,10,11)(H,8,12,13)

InChI key

TZDMCKHDYUDRMB-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Giant vesicle formation through self-assembly of complementary random copolymers.
Ilhan F, et al.
Journal of the American Chemical Society, 122(24), 5895-5896 (2000)
Ojodomo J Achadu et al.
Mikrochimica acta, 185(10), 461-461 (2018-09-17)
A microwave-assisted hydrothermal method was employed to prepare thymine-modified graphitic carbon nitride quantum dots (T-gCNQDs) which are shown to be a novel fluorescent nanoprobe for Hg(II). They exhibit excellent optical properties (blue emission with a fluorescence quantum yield of 46%)
Nan Wang et al.
Biosensors & bioelectronics, 79, 320-326 (2016-01-01)
A sensitive, selective and reusable electrochemical biosensor for the determination of mercury ions (Hg(2+)) has been developed based on thymine (T) modified gold nanoparticles/reduced graphene oxide (AuNPs/rGO) nanocomposites. Graphene oxide (GO) was electrochemically reduced on a glassy carbon substrate. Subsequently
Pilar Amo-Ochoa et al.
Inorganic chemistry, 52(19), 11428-11437 (2013-09-18)
Four new copper(II) coordination complexes, obtained by reaction of CuX2 (X = acetate or chloride) with thymine-1-acetic acid and uracil-1-propionic acid as ligands, of formulas [Cu(TAcO)2(H2O)4]·4H2O (1), [Cu(TAcO)2(H2O)2]n (2), [Cu3(TAcO)4(H2O)2(OH)2]n·4H2O (3), and [Cu3(UPrO)2Cl2(OH)2(H2O)2]n (4) (TAcOH = thymine-1-acetic acid, UPrOH =
Domenica Musumeci et al.
International journal of nanomedicine, 13, 2613-2629 (2018-05-12)
Nucleobase-bearing peptides and their interaction with DNA and RNA are an important topic in the development of therapeutic approaches. On one hand, they are highly effective for modulating the nucleic-acid-based biological processes. On the other hand, they permit to overcome

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service