Skip to Content
Merck
All Photos(2)

Documents

446319

Sigma-Aldrich

Bis(acetonitrile)dichloropalladium(II)

99.99%

Synonym(s):

Palladium(II) chloride diacetonitrile complex

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PdCl2 · (CH3CN)2
CAS Number:
Molecular Weight:
259.43
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

99.99%

form

powder

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

SMILES string

CC#N.CC#N.Cl[Pd]Cl

InChI

1S/2C2H3N.2ClH.Pd/c2*1-2-3;;;/h2*1H3;2*1H;/q;;;;+2/p-2

InChI key

RBYGDVHOECIAFC-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service