Skip to Content
Merck
All Photos(1)

Documents

444006

Sigma-Aldrich

2,2,3,3,4,4,4-Heptafluorobutyl methacrylate

contains MEHQ as inhibitor, 97%

Synonym(s):

HFBMA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=C(CH3)CO2CH2CF2CF2CF3
CAS Number:
Molecular Weight:
268.13
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

liquid

contains

MEHQ as inhibitor

refractive index

n20/D 1.341 (lit.)

bp

134-136 °C (lit.)

density

1.345 g/mL at 25 °C (lit.)

SMILES string

CC(=C)C(=O)OCC(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C8H7F7O2/c1-4(2)5(16)17-3-6(9,10)7(11,12)8(13,14)15/h1,3H2,2H3

InChI key

VIEHKBXCWMMOOU-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

98.6 °F - closed cup

Flash Point(C)

37 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xuanhe Zhang et al.
Nanotechnology, 31(35), 355401-355401 (2020-05-15)
A polymer film capacitor with high energy density has attracted great attention in recent decades to fulfil the requirements of miniaturization and light weight for electronics and energy storage systems. The multi-layered nanocomposite exhibits an effective route for developing polymer
Maciej Jarzębski et al.
Molecules (Basel, Switzerland), 25(9) (2020-04-30)
Fluorescent submicron particles of fluorinated methacrylate (HFMBA) with long-term stability have been synthesized and characterized with regard to their potential applications. Rhodamine B (RBITC) isothiocyanate was used as the fluorescent component. The core-shell structure of the particles effectively protected the
Vishal Vignesh et al.
Biofouling, 37(2), 131-144 (2021-03-19)
Amphiphilic gels consisting of acrylamide (AAM)/2-hydroxyethyl methacrylate (HEMA), hexafluorobutyl methacrylate (HFBMA) and non-isocyanate urethane dimethacrylate (NIUDMA) of varying molecular weights were compared. A three-level Taguchi analysis was performed using the amount of AAM/HEMA, HFBMA, NIUDMA and reaction time as dependent
G Kristin Jonsson et al.
Colloid and polymer science, 295(10), 1983-1991 (2017-10-11)
Fluorinated spheres with grafted poly(ethylene glycol) (PEG) have been synthesized using a semi-batch emulsion polymerization in which the initiator is fed slowly to the reaction. In this way, PEG-grafted colloidal spheres can be fabricated with varying PEG chain length, different

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service