Skip to Content
Merck
All Photos(2)

Key Documents

437417

Sigma-Aldrich

Barium thiosulfate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BaS2O3
CAS Number:
Molecular Weight:
249.46
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22
grade:
for analytical purposes
form:
solid

grade

for analytical purposes

Quality Level

Assay

98%

form

solid

SMILES string

[Ba++].[O-]S([O-])(=O)=S

InChI

1S/Ba.H2O3S2/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

ONPIOWQPHWNPOQ-UHFFFAOYSA-L

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service