Skip to Content
Merck
All Photos(2)

Key Documents

434558

Sigma-Aldrich

Poly(4-styrenesulfonic acid-co-maleic acid) sodium salt

average Mw ~20,000, powder

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(C6H4SO3R)]x[CH(CO2R)CH(CO2R)]y, R=H or Na
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

powder

Quality Level

mol wt

average Mw ~20,000

viscosity

30 cP, 30 % in H2O(lit.)

density

0.94 g/mL at 25 °C

SMILES string

[Na+].O2C(=O)C=CC2=O.c1(ccccc1)C=C

InChI

1S/C8H8.C4H2O3.Na/c1-2-8-6-4-3-5-7-8;5-3-1-2-4(6)7-3;/h2-7H,1H2;1-2H;/q;;+1

InChI key

AGYUYWQDVRLUBV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1:1 4-styrenesulfonic acid:maleic acid mole ratio

Application

Scale control. Dispersant for pigments, clays and fillers.

Features and Benefits

High charge density. Slight surface activity, low foaming.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hugo Vara et al.
Acta biomaterialia, 90, 71-86 (2019-03-25)
Intraspinal microstimulation (ISMS) may help to restore motor functions after spinal cord injury. ISMS caudal to the lesion activates motoneurons and evokes selective movements with graded force in rats and other mammals. We investigated the safety and effectiveness of conducting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service