Skip to Content
Merck
All Photos(1)

Key Documents

430021

Sigma-Aldrich

Linseed Oil

Synonym(s):

Flaxseed oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

liquid

refractive index

n20/D 1.4795

density

0.93 g/mL at 25 °C ()

General description

Linseed oil is a natural triglyceride and a well known drying oil. Drying of the triglyceride mainly leads to the oxidation of the unsaturated fatty acid segments and polymerization. Oxidative degradation studies of linseed oil show hardening and oxidation of alkylic sections followed by partial fragmentation of the triglyceride structure.

Application

Linseed oil filled urea–formaldehyde resin microcapsules may be used to fix cracks in paints/coatings. This linseed oil is processed to create a product that is more suited for many uses (such as paint, to name one), in terms of ″drying time.″

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Drying and oxidative degradation of linseed oil
Lazzari M and Chiantore O
Polymer Degradation and Stability, 65(2), 303-313 (1999)
Drier influence on the curing of linseed oil.
Mallegol J, et al.
Progress in Organic Coatings, 39(2), 107-113 (2000)
Preparation and characterization of microcapsules containing linseed oil and its use in self-healing coatings.
Suryanarayana CK, et al.
Progress in Organic Coatings, 63(1), 72-78 (2008)
Pyrolysis of pine wood in a slowly heating fixed-bed reactor: Potassium carbonate versus calcium hydroxide as a catalyst
Wang Z, et al.
Fuel Processing Technology, 91(8), 942-950 (2010)
Wan Maznah Wan Ishak et al.
Drug delivery and translational research, 9(2), 418-433 (2018-04-19)
Wound healing is a physiological event that generates reconstitution and restoration of granulation tissue that ends with scar formation. As omega fatty acids are part of membrane phospholipids and participate in the inflammatory response, we investigated the effects of omega-3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service