Skip to Content
Merck
All Photos(3)

Key Documents

421804

Sigma-Aldrich

(1S,2S)-trans-1,2-Cyclohexanediol

99%

Synonym(s):

(1S)-trans-1,2-Cyclohexanediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H10(OH)2
CAS Number:
Molecular Weight:
116.16
Beilstein:
1901343
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

optical activity

[α]20/D +39°, c = 1.6 in H2O

optical purity

ee: 99% (GLC)

mp

107-109 °C (lit.)

SMILES string

O[C@H]1CCCC[C@@H]1O

InChI

1S/C6H12O2/c7-5-3-1-2-4-6(5)8/h5-8H,1-4H2/t5-,6-/m0/s1

InChI key

PFURGBBHAOXLIO-WDSKDSINSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(1S,2S)-trans-1,2-Cyclohexanediol may be used in the preparation of (1S,2S)-1,2-cyclohexanediyl bis(4-vinylbenzoate) by reacting with 4-vinylbenzoyl chloride. It may also be used as a chiral ligand for the preparation of non-racemic hydroxy phosphonates via titanium alkoxide-catalyzed addition of dimethyl phosphite to the corresponding aldehydes.
C2-symmetric chiral diol with versatile applications as a chiral auxiliary, building block, and chiral ligand.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chirality induction in cyclocopolymerization. 14. Template effect of 1, 2-cycloalkanediol in the cyclocopolymerization of bis (4-vinylbenzoate) s with styrene.
Kakuchi T, et al.
Macromolecules, 34(1), 38-43 (2001)
The synthesis of 1-hydroxy phosphonates of high enantiomeric excess using sequential asymmetric reactions: titanium alkoxide-catalyzed P C bond formation and kinetic resolution.
Rowe BJ and Spilling CD.
Tetrahedron Asymmetry, 12(12), 1701-1708 (2001)
Burk, M.J. et al.
Tetrahedron Asymmetry, 2, 569-569 (1991)
Ho, O.C. et al.
Organometallics, 14, 2855-2855 (1995)
Marshall, J.A. Xie, S.
The Journal of Organic Chemistry, 60, 7230-7230 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service