Skip to Content
Merck
All Photos(1)

Key Documents

421014

Sigma-Aldrich

Bis[2-(2-butoxyethoxy)ethyl] adipate

Synonym(s):

BBOEA, Di(butoxyethoxyethyl) adipate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2CH2CO2(CH2CH2O)2(CH2)3CH3]2
CAS Number:
Molecular Weight:
434.56
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

liquid

impurities

<1% diethylene glycol monobutyl ether

refractive index

n20/D 1.448 (lit.)

mp

−11 °C (lit.)

density

1.01 g/mL at 25 °C (lit.)

SMILES string

CCCCOCCOCCOC(=O)CCCCC(=O)OCCOCCOCCCC

InChI

1S/C22H42O8/c1-3-5-11-25-13-15-27-17-19-29-21(23)9-7-8-10-22(24)30-20-18-28-16-14-26-12-6-4-2/h3-20H2,1-2H3

InChI key

SCABKEBYDRTODC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Plasticizer

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

410.0 °F - open cup

Flash Point(C)

210 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Do Young Kim et al.
Polymers, 12(9) (2020-08-28)
The mechanical properties and tear resistance of an ecofriendly flexible packaging film, i.e., poly(lactic acid) (PLA)/poly (butylene adipate-co-terephthalate) (PBAT) film, were investigated via a blown film extrusion process. The application of PLA and PBAT in product packaging is limited due

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service