Skip to Content
Merck
All Photos(1)

Key Documents

41571

Sigma-Aldrich

1,1-Dimethylsiletane

≥95%

Synonym(s):

1,1-Dimethylsilacyclobutane, Dimethyltrimethylenesilane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12Si
CAS Number:
Molecular Weight:
100.23
Beilstein:
102434
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

form

liquid

refractive index

n20/D 1.428

density

0.777 g/mL at 20 °C (lit.)

SMILES string

C[Si]1(C)CCC1

InChI

1S/C5H12Si/c1-6(2)4-3-5-6/h3-5H2,1-2H3

InChI key

YQQFFTNDQFUNHB-UHFFFAOYSA-N

Other Notes

Reactive silicon compound undergoing various insertion reactions giving 5- and 6-membered rings

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

23.0 °F - closed cup

Flash Point(C)

-5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K. Matsumoto et al.
Tetrahedron Letters, 31, 6055-6055 (1990)
Tetrahedron Letters, 31, 6059-6059 (1990)
Y. Kakeyama et al.
Bulletin of the Chemical Society of Japan, 64, 1461-1461 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service