Skip to Content
Merck
All Photos(3)

Key Documents

409693

Sigma-Aldrich

Octadecyl acrylate

contains 200 ppm monomethyl ether hydroquinone as inhibitor, 97%

Synonym(s):

Stearyl acrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCO2(CH2)17CH3
CAS Number:
Molecular Weight:
324.54
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

97%

form

solid

contains

200 ppm monomethyl ether hydroquinone as inhibitor

mp

32-34 °C (lit.)

density

0.8 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCCOC(=O)C=C

InChI

1S/C21H40O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-23-21(22)4-2/h4H,2-3,5-20H2,1H3

InChI key

FSAJWMJJORKPKS-UHFFFAOYSA-N

Application

Octadecyl acrylate undergoes surface-initiated atom transfer radical polymerization (ATRP) to form poly(octadecyl acrylate). ODA can form copolymers with cinnamoyloxy ethyl methacrylate (CEMA).

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crosslinked reactive macromonomers based on polyisobutylene and octadecyl acrylate copolymers as crude oil sorbers.
Atta AM, et al.
Reactive functional Polymers, 66(9), 931-943 (2006)
Facile synthesis of high-density poly (octadecyl acrylate)-grafted silica for reversed-phase high-performance liquid chromatography by surface-initiated atom transfer radical polymerization.
Mallik AK, et al.
Journal of Chromatography A, 1187(1), 119-127 (2008)
Yusuke Akino et al.
Polymers, 11(2) (2019-04-10)
The effect of hydration in corona layer on temperature responsiveness of polymer micelles consisting of poly(N-vinyl pyrrolidone)-block-poly(n-octadecyl acrylate) (PVP-b-PODA) was investigated. Small-angle X-ray scattering and dynamic light scattering showed two-step shape change of PVP-b-PODA micelles around 45 and 65 °C
Gordana Siljanovska Petreska et al.
Polymers, 11(8) (2019-08-02)
High molecular weight waterborne ABA block copolymers of styrene (St) and 2-ethylhexyl acrylate (2EHA) containing hard and soft domains were synthesized by means of RAFT (mini)emulsion polymerization using a bifunctional symmetric S,S-dibenzyl trithiocarbonate (DBTTC) RAFT agent. Miniemulsion polymerization was initially
Gordana Siljanovska Petreska et al.
Polymers, 12(6) (2020-06-04)
AB diblock waterborne copolymers made of styrene (St) and 2-ethylhexyl acrylate (2EHA) were synthesized by means of two-step reversible addition fragmentation chain transfer (RAFT) (mini)emulsion polymerization. Monofunctional asymmetric RAFT agent was used to initiate the polymerization. The hard polystyrene "A"

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service