Skip to Content
Merck
All Photos(1)

Key Documents

409081

Sigma-Aldrich

Poly(acrylamide-co-diallyldimethylammonium chloride) solution

10 wt. % in H2O

Synonym(s):

P(AAm-co-DADMAC)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

viscous liquid

composition

acrylamide, ~55 wt. %

concentration

10 wt. % in H2O

viscosity

9,000-25,000 cP(25 °C, Brookfield)(lit.)

density

1.02 g/mL at 25 °C

SMILES string

Cl.[N+](CC=C)(CC=C)(C)C.N=C([O-])C=C

InChI

1S/C8H16N.C3H5NO.ClH/c1-5-7-9(3,4)8-6-2;1-2-3(4)5;/h5-6H,1-2,7-8H2,3-4H3;2H,1H2,(H2,4,5);1H/q+1;;/p-1

InChI key

XFOSBZOUUACCCN-UHFFFAOYSA-M

Application

Waste treatment for laundry, emulsion breaking, sludge dewatering and drainage and retention aid. Cationic polyelectrolyte

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gloria Sánchez-Galván et al.
World journal of microbiology & biotechnology, 35(1), 14-14 (2019-01-04)
The effects of bicarbonate loading rate (BLR) and pH on growth kinetics, inorganic carbon speciation, carbon fixation and lipid content in Neochloris oleoabundans cultures using anaerobically digested stillage (ADS) (2% v/v) were investigated. Four different cultures were established: culture A
Adél Szerlauth et al.
Soft matter, 16(46), 10518-10527 (2020-10-20)
Highly stable antioxidant dispersions were designed on the basis of ring-opened ellagic acid (EA) intercalated into MgAl-layered double hydroxide (LDH) nanoparticles. The morphology of the composite was delicately modified with ethanolic washing to obtain EtOH-EA-LDH with a high specific surface
Allergic contact dermatitis from laureth-9 and polyquaternium-7 in a skin-care product.
R Gallo et al.
Contact dermatitis, 45(6), 356-357 (2002-02-16)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service