Skip to Content
Merck
All Photos(2)

Documents

403105

Sigma-Aldrich

Poly(3-hydroxybutyric acid-co-3-hydroxyvaleric acid)

natural origin, PHV content 8 mol %

Synonym(s):

PHBV

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[COCH2CH(CH3)O]m[COCH2CH(C2H5)O]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

description

8 mol% is approximately 9 wt%.

Quality Level

composition

PHV content, 8 mol %

SMILES string

CC(O)CC(O)=O.CCC(O)CC(O)=O

InChI

1S/C5H10O3.C4H8O3/c1-2-4(6)3-5(7)8;1-3(5)2-4(6)7/h4,6H,2-3H2,1H3,(H,7,8);3,5H,2H2,1H3,(H,6,7)

InChI key

IUPHTVOTTBREAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Poly(3-Hydroxybutyrate-co-3-Hydroxyvalerate): Enhancement Strategies for Advanced Applications: Review on enhancing PHBV for advanced applications, discussing various strategies for improving its properties (Rivera-Briso & Serrano-Aroca, 2018).

  • In vitro characterization and nuclear delivery of poly (3-hydroxybutyric acid-co-3-hydroxyvaleric acid)(PHBV) based nanoparticles: Preparation and study of PHBV nanocapsules for penetration into human osteosarcoma and L929 cells (Şahin, 2015).

Features and Benefits

Biodegradable polymer

Preparation Note

Produced via a controlled fermentation process using microoroganisms.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service