401404
Cobalt(II) perchlorate hexahydrate
Synonym(s):
Cobaltous perchlorate hexahydrate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
Co(ClO4)2 · 6H2O
CAS Number:
Molecular Weight:
365.93
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
crystals and lumps
Recommended Products
form
crystals and lumps
Quality Level
concentration
13.8-16.9% Co (EDTA titration)
application(s)
battery manufacturing
SMILES string
O.O.O.O.O.O.[Co++].[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O
InChI
1S/2ClHO4.Co.6H2O/c2*2-1(3,4)5;;;;;;;/h2*(H,2,3,4,5);;6*1H2/q;;+2;;;;;;/p-2
InChI key
KPOBHNYTWJSVKF-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 2 - Ox. Sol. 2
Storage Class Code
5.1A - Strongly oxidizing hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Magdalena Woźniczka et al.
Journal of inorganic biochemistry, 187, 62-72 (2018-07-29)
The coordination properties of 2-picolinehydroxamic acid towards cobalt(II) in aqueous solution were determined by a pH-metric method and confirmed by spectroscopic (UV-Vis and ESI-MS) studies. The results show the formation of mononuclear complexes, as well as of metallacrowns (MC). All
Magdalena Woźniczka et al.
Molecules (Basel, Switzerland), 25(15) (2020-08-06)
The present study describes the coordination properties of a reduced Schiff base, N-(2-hydroxybenzyl)alanine, towards cobalt(II) using potentiometric as well as spectroscopic (UV-Vis and ESI-MS) methods. The results indicate the formation of six mononuclear complexes showing high stability in aqueous solution.
Greg Brewer et al.
Materials (Basel, Switzerland), 13(7) (2020-04-05)
The 3:1 condensation of 5-methyl-1H-pyrazole-3-carboxaldehyde (MepyrzH) with tris(2-aminoethyl)amine (tren) gives the tripodal ligand tren(MePyrzH)3. Aerial oxidation of a solution of cobalt(II) with this ligand in the presence of base results in the isolation of the insoluble Co(tren)(MePyrz)3. This complex reacts
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service