Skip to Content
Merck
All Photos(1)

Key Documents

400874

Sigma-Aldrich

Iron(II) titanate

−100 mesh, 99.9%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FeTiO3
CAS Number:
Molecular Weight:
151.71
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9%

form

powder

particle size

−100 mesh

SMILES string

[Fe++].[O-][Ti]([O-])=O

InChI

1S/Fe.3O.Ti/q+2;;2*-1;

InChI key

WIBXORLWTCPWFI-UHFFFAOYSA-N

Related Categories

General description

Iron(II) titanate is a mixed metal oxide known for its unique magnetic and electrical properties, which are influenced by its orthorhombic or monoclinic crystal structure. It is widely used in photocatalysis due to its excellent photocatalytic activity. Additionally, Iron(II) titanate is utilized in lithium-ion batteries as an electrode material, enhancing the battery′s performance and longevity.

Application

Iron(II) titanate is utilized as a photocatalyst in solar energy conversion processes, particularly for water splitting and hydrogen production. Its ability to absorb visible light makes it suitable for enhancing photocatalytic efficiency. FeTiO3 is explored as a material for electrodes in batteries and supercapacitors. Its unique properties contribute to improved charge storage and conductivity, enhancing the performance of energy storage devices.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service