Skip to Content
Merck
All Photos(2)

Key Documents

400645

Sigma-Aldrich

Silver(I) telluride

greener alternative
Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ag2Te
CAS Number:
Molecular Weight:
343.34
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

form

powder and chunks

Quality Level

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

density

8.5 g/mL at 25 °C (lit.)

greener alternative category

SMILES string

[Ag][Te][Ag]

InChI

1S/2Ag.Te

InChI key

YRXWPCFZBSHSAU-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Silver(I) telluride (Ag2Te) is a silver ion conductor, which can be used as a thermoelectric material for applications in power generation from waste heat. It can be used in the formation of p-type conductor for the fabrication of semiconducting devices.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phase Transition of Ag-Enriched Ag2 Te
Aliev FF, et al.
Inorganic Chemistry, 38(10), 995- 997 null
Preparation and thermoelectric properties of sulfur doped Ag 2 Te nanoparticles via solvothermal methods
Zhou W, et al.
Nanoscale, 4(13), 3926-3931 (2012)
Effect of phase transition on the thermoelectric properties of Ag2Te
Jung D, et al.
Materials Transactions, 1216-1219 (2012)
Synergism in binary nanocrystal superlattices leads to enhanced p-type conductivity in self-assembled PbTe/Ag 2 Te thin films
Urban JJ, et al.
Testing, 6(2), 115-121 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service