Skip to Content
Merck
All Photos(1)

Key Documents

395102

Sigma-Aldrich

Tris(4-methoxyphenyl)phosphine

95%

Synonym(s):

TPAP, Tris(p-anisyl)phosphine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3OC6H4)3P
CAS Number:
Molecular Weight:
352.36
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand
reaction type: Cross Couplings

mp

131-134 °C (lit.)

functional group

phosphine

SMILES string

COc1ccc(cc1)P(c2ccc(OC)cc2)c3ccc(OC)cc3

InChI

1S/C21H21O3P/c1-22-16-4-10-19(11-5-16)25(20-12-6-17(23-2)7-13-20)21-14-8-18(24-3)9-15-21/h4-15H,1-3H3

InChI key

UYUUAUOYLFIRJG-UHFFFAOYSA-N

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haigang Wu et al.
Scientific reports, 7, 41299-41299 (2017-02-02)
Doping via self-assembled macromolecules might offer a solution for developing single atom electronics by precisely placing individual dopants at arbitrary location to meet the requirement for circuit design. Here we synthesize dendrimer-like polyglycerol macromolecules with each carrying one phosphorus atom
T Srinivasa Reddy et al.
Dalton transactions (Cambridge, England : 2003), 47(43), 15312-15323 (2018-09-07)
A series of gold(i), gold(iii) and cationic gold(i) complexes of tris(4-methoxyphenyl)phosphine and tris(2,6-dimethoxyphenyl)phosphine were synthesised and fully characterised by spectroscopic methods. The molecular structures of selected complexes were also determined by X-ray diffraction analysis. The prepared complexes [AuX{P(C6H4-4-OMe)3}] [X =

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service