393665
1,1-Dibromo-2,2-bis(chloromethyl)cyclopropane
90%, technical grade
Synonym(s):
1,1-Dibromo-2,2-bis(chloromethyl)cyclopropane
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
grade
technical grade
Quality Level
Assay
90%
form
solid
mp
48-50 °C (lit.)
functional group
bromo
chloro
SMILES string
ClCC1(CCl)CC1(Br)Br
InChI
1S/C5H6Br2Cl2/c6-5(7)1-4(5,2-8)3-9/h1-3H2
InChI key
RCRVZCIUKQNOIS-UHFFFAOYSA-N
Related Categories
General description
1,1-Dibromo-2,2-bis(chloromethyl)cyclopropane is a halogenated cyclopropane derivative. It has been reported to be synthesized from 3-chloro-2-(chloromethyl)-1-propene. It is formed as an intermediate during the synthesis of [1.1.1]propellane by Szeimies method.
Application
1,1-Dibromo-2,2-bis(chloromethyl)cyclopropane may be used as a starting material in the synthesis of [1.1.1]-propellane and ((3-ethynylbicyclo[1.1.1]pentan-1-yl)ethynyl)trimethylsilane.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Journal of the American Chemical Society, 135(25), 9366-9376 (2013-06-04)
The rod-like molecule bis((4-(4-pyridyl)ethynyl)bicyclo[2.2.2]oct-1-yl)buta-1,3-diyne, 1, contains two 1,4-bis(ethynyl)bicyclo[2.2.2]octane (BCO) chiral rotators linked by a diyne fragment and self-assembles in a one-dimensional, monoclinic C2/c centrosymmetric structure where two equilibrium positions with large occupancy imbalance (88% versus 12%) are identified on a
Improved Preparations of 3-Chloro-2-(chloromethyl)-1-propene and 1, 1-Dibromo-2, 2-bis (chloromethyl) cyclopropane: Intermediates in the Synthesis of [1.1. 1] Propellane.
The Journal of Organic Chemistry, 60(14), 4666-4668 (1995)
[1.1. 1] Propellane: Reaction with Singlet Dihalocarbene.
Bull. Korean Chem. Soc., 19(3), 367-369 (1998)
Concerning the synthesis of [1.1. 1] propellane.
Chemische Berichte, 122(2), 397-398 (1989)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service