All Photos(1)
About This Item
Linear Formula:
C6H4(COC6H5)2
CAS Number:
Molecular Weight:
286.32
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
solid
mp
105-108 °C (lit.)
functional group
ketone
phenyl
SMILES string
O=C(c1ccccc1)c2cccc(c2)C(=O)c3ccccc3
InChI
1S/C20H14O2/c21-19(15-8-3-1-4-9-15)17-12-7-13-18(14-17)20(22)16-10-5-2-6-11-16/h1-14H
InChI key
MJQHDSIEDGPFAM-UHFFFAOYSA-N
General description
1,3-Dibenzoylbenzene ([3-(benzoyl)phenyl]-phenyl-methanone, m-Dibenzoylbenzene) is a diarylketone derivative. It has topologically pseudogenerate Π-LUMO′s (lowest unoccupied molecular orbital). Its synthesis from benzene has been reported. Crystal structure studies suggest that its crystals are orthorhombic and exhibit space goup Pbca.
Application
1,3-Dibenzoylbenzene ([3-(benzoyl)phenyl]-phenyl-methanone, m-Dibenzoylbenzene) is suitable reagent used in the synthesis of 1,3-bis(1-phenylvinyl)benzene (MDDPE). It may be used in the synthesis of polyanionic spin-quintet molecular clusters and ground state undecet hydrocarbon with 10 parallel spins.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Synthesis and characterization of ABC-type star and linear block copolymers of styrene, isoprene, and 1, 3-cyclohexadiene.
Wang X, et al.
Macromolecules, 39(20), 6898-6904 (2006)
Design, preparation, and electron spin resonance detection of a ground-state undecet (S= 5) hydrocarbon.
Fujita I, et al.
Journal of the American Chemical Society, 112(10), 4074-4075 (1990)
(3-Benzoylphenyl)(phenyl) methanone.
Raza Ahsraf A, et al.
Acta Crystallographica Section E, Structure Reports Online, 67(9), 2432-2432 (2001)
Shigeaki Nakazawa et al.
Physical chemistry chemical physics : PCCP, 13(4), 1424-1433 (2010-11-26)
Trianionic spin-quartet and tetraanionic spin-quintet molecular clusters derived from m-dibenzoylbenzene in solution were identified by CW-ESR/pulse-ESR based two-dimensional electron spin transient nutation spectroscopy, and their spin and clustering structures in the ground state were determined in terms of a D-tensor
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service