Skip to Content
Merck
All Photos(1)

Key Documents

388866

Sigma-Aldrich

Brij® 93

average Mn ~357

Synonym(s):

Polyethylene glycol oleyl ether, Polyoxyethylene (2) oleyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C18H35(OCH2CH2)nOH, n~2
CAS Number:
Molecular Weight:
356.58
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23
Pricing and availability is not currently available.

vapor pressure

0.26 psi ( 20 °C)

form

liquid

mol wt

average Mn ~357

refractive index

n20/D 1.462 (lit.)

density

0.912 g/mL at 25 °C (lit.)

HLB

4

SMILES string

O(CCO)CCCCCCCC\C=C/CCCCCCCC

InChI

1S/C20H40O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-19-22-20-18-21/h9-10,21H,2-8,11-20H2,1H3/b10-9-

InChI key

KWVPFECTOKLOBL-KTKRTIGZSA-N

General description

Brij® 93 is a non-ionic surfactant which has a polar group in the form of polyoxyethylene that is separated by the polyethylene chains. Its molecular structure includes polyethylene glycol (PEG) single chain. It is slightly soluble in water and has a hydrophobic nature.[1][2][3]

Legal Information

Brij is a registered trademark of Croda International PLC

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

>464.0 °F - Equilibrium method

Flash Point(C)

> 240 °C - Equilibrium method

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stabilization of High Ionic Strength Slurries Using Surfactant Mixtures: Molecular Factors That Determine Optimal Stability
Palla BJ, et al.
Journal of Colloid and Interface Science, 256(1), 143-152 (2002)
Optimization of a novel and improved thermosensitive liposome formulated with DPPC and a Brij surfactant using a robust in vitro system
Tagami T, et al.
J. Controlled Release, 154(3), 290-297 (2011)
Hydrothermal synthesis of nanocrystalline hydroxyapatite from phosphogypsum waste
Bensalah H, et al.
Journal of Environmental Chemical Engineering, 6(1), 1347-1352 (2018)
Yash Kapoor et al.
International journal of pharmaceutics, 361(1-2), 222-229 (2008-06-26)
Cyclosporine A (CyA) is an immunosuppressant drug that is used for treating a variety of ocular diseases and disorders. CyA is commonly delivered via eye drops, which is highly inefficient due to a low bioavailability of less than 5%. The
R B Shah et al.
International journal of pharmaceutics, 341(1-2), 189-194 (2007-05-25)
In the present work, a novel application of ultrasonic measurements is detailed to characterize nano-emulsion formulations as a part of the overall Quality by Design (QbD) goal. Ultrasonic resonator technology (URT) was utilized to measure sound velocity and absorption of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service