388726
Polybutenes
average Mn ~2,300 by VPO, isobutylene >90 %
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
[-CH2C(CH3)2-]x[-CH2CH(C2H5)-]y
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
form
liquid
mol wt
average Mn ~2,300 by VPO
composition
isobutylene, >90%
refractive index
n20/D 1.504
viscosity
4,000-4,400 cSt(99 °C)(lit.)
transition temp
Tg −64.9 °C
pour point (ASTM D 97) +18 °C
density
0.908 g/mL at 25 °C
SMILES string
CCC=C.C\C=C\C
InChI
1S/2C4H8/c2*1-3-4-2/h3-4H,1-2H3;3H,1,4H2,2H3/b4-3+;
InChI key
WTOOLIQYCQJDBG-BJILWQEISA-N
Looking for similar products? Visit Product Comparison Guide
Application
Hydrophobic material is derivatized to make detergents and anti-rust additives. Adds tack, flexibility and water repellency to adhesives and coatings. Contributes ′cling′ to LLDPE films. Plasticizer for polymers, lubricants and metal-working fluids.
Features and Benefits
Tackiness increases with molecular weight. Some internal double bonds may be present.
Physical form
Terminal vinyl group.
Other Notes
remainder 1- and 2-butenes
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Asp. Tox. 1 - Skin Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
159.8 °F - closed cup
Flash Point(C)
71 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
X Y Wu et al.
The British journal of ophthalmology, 82(5), 569-576 (1998-08-26)
The goal was to compare the biological response of the corneal stroma with three porous materials: a melt blown microfibre web of polybutylene:polypropylene (80:20); a polyester spun laced fabric (polyethylene terephthalate), and an expanded polytetrafluoroethylene. Fifty per cent of each
Huaiyu Wang et al.
Acta biomaterialia, 5(1), 279-287 (2008-09-02)
Poly(butylene succinate) (PBSu), a novel biodegradable aliphatic polyester with excellent processability and mechanical properties, is a promising substance for bone and cartilage repair. However, it typically suffers from insufficient biocompatibility and bioactivity after implantation into the human body. In this
X Guo et al.
Guang pu xue yu guang pu fen xi = Guang pu, 21(2), 169-172 (2003-09-02)
Hydrogen bonding in hard segment domains of segmented polyester polyurethaneurea (PUU) based on 4,4'-diphenylmethane diisocyanate (MDI), ethylene diamine (ED) and polybutylene adipate (PBA) (M.W.1,975 and M.W.1,228) was studied by means of IR spectroscopy. It was found that the double peak
Syutaro Yamamoto et al.
Journal of oleo science, 56(9), 463-469 (2007-09-28)
Water-soluble metal working fluids are used for processing of aluminum alloy materials. This short article describes properties of new additives in water-soluble metal working fluids for aluminum alloy materials. Many half esters or diesters were prepared from the reactions of
A M Belu et al.
Rapid communications in mass spectrometry : RCM, 14(7), 564-571 (2000-04-25)
A series of 16 polyarylates, with well-controlled and systematically varying chemistry, has been characterized by time-of-flight secondary ion mass spectrometry (TOF-SIMS). The polymers are structurally identical except for the incremental additions of C2H4 units to the backbone and sidechain. From
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service