Skip to Content
Merck
All Photos(1)

Key Documents

383163

Sigma-Aldrich

Niobium(IV) oxide

99.9% trace metals basis

Synonym(s):

Niobium dioxide, Niobium dioxide (NbO2 ), Niobium oxide (Nb2 O4 )

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NbO2
CAS Number:
Molecular Weight:
124.91
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% trace metals basis

form

powder

density

5.9 g/mL at 25 °C (lit.)

SMILES string

O=[Nb]=O

InChI

1S/Nb.2O

InChI key

HFLAMWCKUFHSAZ-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service