Skip to Content
Merck
All Photos(3)

Key Documents

382310

Sigma-Aldrich

Didecyldimethylammonium bromide

98%

Synonym(s):

DDAB

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)9]2N(CH3)2Br
CAS Number:
Molecular Weight:
406.53
Beilstein:
6492993
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

149-151 °C (lit.)

SMILES string

[Br-].CCCCCCCCCC[N+](C)(C)CCCCCCCCCC

InChI

1S/C22H48N.BrH/c1-5-7-9-11-13-15-17-19-21-23(3,4)22-20-18-16-14-12-10-8-6-2;/h5-22H2,1-4H3;1H/q+1;/p-1

InChI key

UMGXUWVIJIQANV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Didecyldimethylammonium bromide (DDAB) is commonly used as a surfactant and a phase transfer catalyst. It is also used in the preparation of hydrophobic ammonium-based ionic liquid.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jie Qu et al.
Drug delivery, 23(9), 3408-3416 (2016-05-18)
Glioblastoma is a malignant brain tumor originating in the central nervous system. Successfully therapy of this disease required the efficient delivery of therapeutic agents to the tumor cells and tissues. Delivery of anticancer drugs using novel nanocarriers is promising in
Copper?Metallomesogen Structures Obtained by Ionic Self?Assembly (ISA): Molecular Electromechanical Switching Driven by Cooperativity.
Camerel F, et al.
Chemistry?A European Journal , 9(16), 3764-3771 (2003)
Ruthenium (III)-catalysed hydrogen peroxide oxidation of alkyl aromatic compounds under phase-transfer conditions.
Barak G and Sasson Y
Journal of the Chemical Society, 10, 637-638 (1988)
Long alkyl chain quaternary ammonium-based ionic liquids and potential applications.
Pernak J, et al.
Green Chemistry, 8(9), 798-806 (2006)
Regis Marty et al.
Nucleic acids research, 37(15), 5197-5207 (2009-06-30)
Despite considerable interest and investigations on cationic lipid-DNA complexes, reports on lipid-RNA interaction are very limited. In contrast to lipid-DNA complexes where lipid binding induces partial B to A and B to C conformational changes, lipid-tRNA complexation preserves tRNA folded

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service