Skip to Content
Merck
All Photos(1)

Key Documents

381241

Sigma-Aldrich

Diethylphenylphosphine

96%

Synonym(s):

NSC 158475

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2PC6H5
CAS Number:
Molecular Weight:
166.20
Beilstein:
742484
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

96%

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

refractive index

n20/D 1.546 (lit.)

bp

120-121 °C/29 mmHg (lit.)

density

0.954 g/mL at 25 °C (lit.)

functional group

phosphine

SMILES string

CCP(CC)c1ccccc1

InChI

1S/C10H15P/c1-3-11(4-2)10-8-6-5-7-9-10/h5-9H,3-4H2,1-2H3

InChI key

LVTCZSBUROAWTE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Catalyst for:
  • Selective cross-dimerization
  • Diastereoselective condensations
  • Carboxyl migration reactions
  • Selective hydrogenation
  • Asymmetric induction by chiral diphosphines in ring contraction

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

183.2 °F - closed cup

Flash Point(C)

84 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B P Smyser et al.
Biochemical pharmacology, 35(10), 1719-1723 (1986-05-15)
Purified mouse liver cytochrome P-450 reconstituted with purified NADPH-cytochrome P-450 reductase and phosphatidylcholine metabolized diethylphenylphosphine to diethylphenylphosphine oxide. NADPH was required for the reaction and the amount of oxide formed was time and cytochrome P-450 dependent. Purified phenobarbital-induced cytochrome P-450

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service