Skip to Content
Merck
All Photos(1)

Key Documents

379212

Sigma-Aldrich

Hexamethyldisilazane

Hexamethyldisilazane

ReagentPlus®, 99.9%

Synonym(s):

1,1,1,3,3,3-Hexamethyldisilazane, HMDS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiNHSi(CH3)3
CAS Number:
Molecular Weight:
161.39
Beilstein:
635752
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

product line

ReagentPlus®

Assay

99.9%

form

liquid

refractive index

n20/D 1.407 (lit.)

bp

125 °C (lit.)

SMILES string

C[Si](C)(C)N[Si](C)(C)C

InChI

1S/C6H19NSi2/c1-8(2,3)7-9(4,5)6/h7H,1-6H3

InChI key

FFUAGWLWBBFQJT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hexamethyldisilazane (HDMS) is an organoslicon compound. HDMS is a colorless liquid used as a reagent and precursor in organic synthesis and organometallic chemistry. It is also used as a sol-gel precursor to fabricate thin films for semiconductor devices.[1] [2]

Application

Hexamethyldisilazane can be used:
  • As a precursor to fabricate multilayer quantum dot light-emitting diodes.[1]
  • To modify dielectric surfaces of organic thin film transistors.[3]
  • To fabricate superhydrophobic coatings on epoxy resins.[4]
  • For the silylation of alcohols and phenols.[2]

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

52.5 °F - closed cup

Flash Point(C)

11.4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I-Shuo Huang et al.
Biology, 9(8) (2020-07-28)
Aside from two samples collected nearly 50 years ago, little is known about the microbial composition of wind tidal flats in the hypersaline Laguna Madre, Texas. These mats account for ~42% of the lagoon's area. These microbial communities were sampled
G Chiavari et al.
Journal of chromatography. A, 922(1-2), 235-241 (2001-08-07)
Alpha-amino acids were pyrolysed at 600 degrees C in the presence of hexamethyldisilazane (HMDS) and the formed volatile products were analysed on line by gas chromatography-mass spectrometry (GC-MS). Glycine, alanine, valine, leucine, isoleucine, norleucine, methionine, phenylalanine yielded principally the trimethylsilyl
A simple and efficient room temperature silylation of diverse functional groups with hexamethyldisilazane using CeO2 nanoparticles as solid catalysts
Nagaraj Anbu, et al
Molecular Catalysis, 474, 110357-110357 (2019)
High performance organic thin film transistors using chemically modified bottom contacts and dielectric surfaces
Organic Electronics, 15, 2073-2078 (2014)
Joachim C Burbiel et al.
Beilstein journal of organic chemistry, 2, 20-20 (2006-10-28)
Poly-substituted xanthine derivatives are an important class of compounds in medicinal chemistry. Substitution at the 8-position of the purine ring is generally accessible by ring closure of a carboxamido-substituted uracil precursor. Although several procedures to accomplish this synthetic step have

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service