Skip to Content
Merck
All Photos(1)

Key Documents

376973

Sigma-Aldrich

9,9-Dimethylxanthene

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14O
CAS Number:
Molecular Weight:
210.27
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

bp

114-115 °C/0.6 mmHg (lit.)

mp

35-38 °C (lit.)

SMILES string

CC1(C)c2ccccc2Oc3ccccc13

InChI

1S/C15H14O/c1-15(2)11-7-3-5-9-13(11)16-14-10-6-4-8-12(14)15/h3-10H,1-2H3

InChI key

MTVNAPYHLASOSX-UHFFFAOYSA-N

General description

9,9-Dimethylxanthene is a heterocyclic building block.

Application

9,9-Dimethylxanthene is suitable for use in the preparation of 9,9-dimethylxanthene-4,5-dicarboxylic acid. It may be used in the preparation of 9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene. It may be used in the preparation of 4,6-bis(dimesitylboryl)dibenzofuran.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y M Dunayevskiy et al.
Proceedings of the National Academy of Sciences of the United States of America, 93(12), 6152-6157 (1996-06-11)
By means of capillary electrophoresis coupled online to electrospray ionization MS, a library of theoretically 171 disubstituted xanthene derivatives was analyzed. The method allowed the purity and makeup of the library to be determined: 160 of the expected compounds were
Synthesis, Structure, and Cyclic Voltammetry of 4, 6-Bis (dimesitylboryl) dibenzofuran: Isolation of 4, 6-Dilithiobenzofuran and 4, 5-Dilithio-9, 9-dimethylxanthene as tmeda Adducts.
Wang H and Gabbai FP.
Organometallics, 24(12), 2898-2902 (2005)
Bidentate phosphines of heteroarenes: 9, 9-dimethyl-4, 5-bis (diphenylphosphino) xanthene.
Hillebrand S, et al.
Tetrahedron Letters, 36(1), 75-78 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service